missing translation for 'onlineSavingsMsg'
Learn More

1,3,3-Trimethyl-2-methyleneindoline 96.0+%, TCI America™

Catalog No. T087925ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T087925ML 25 mL
T0879100ML 100 mL
T0879500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T087925ML Supplier TCI America Supplier No. T087925ML

Chemical Identifiers

CAS 118-12-7
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.259
MDL Number MFCD00005813
InChI Key ZTUKGBOUHWYFGC-UHFFFAOYSA-N
Synonym 1,3,3-trimethyl-2-methyleneindoline, fischer's base, fischer base, 1h-indole, 2,3-dihydro-1,3,3-trimethyl-2-methylene, fischer's methylene base, 2-methylene-1,3,3-trimethylindoline, fischers base, ccris 6608, indoline, 2-methylene-1,3,3-trimethyl, indoline, 1,3,3-trimethyl-2-methylene
PubChem CID 8351
IUPAC Name 1,3,3-trimethyl-2-methylideneindole
SMILES CC1(C(=C)N(C2=CC=CC=C21)C)C

Specifications

Melting Point -10°C
Color Red
Boiling Point 248°C
Quantity 25 mL
UN Number 3082
Formula Weight 173.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,3-Trimethyl-2-methyleneindoline
RTECSNumber NM1926500
TSCA Yes