missing translation for 'onlineSavingsMsg'
Learn More

1,3,3-Trimethyl-2-methyleneindoline, 95%

Catalog No. AAA1674118
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1674118 50 g
AAA1674130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1674118 Supplier Thermo Scientific Chemicals Supplier No. A1674118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 118-12-7 | C12H15N | 173.259 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-12-7
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.259
MDL Number MFCD00005813
InChI Key ZTUKGBOUHWYFGC-UHFFFAOYSA-N
Synonym 1,3,3-trimethyl-2-methyleneindoline, fischer's base, fischer base, 1h-indole, 2,3-dihydro-1,3,3-trimethyl-2-methylene, fischer's methylene base, 2-methylene-1,3,3-trimethylindoline, fischers base, ccris 6608, indoline, 2-methylene-1,3,3-trimethyl, indoline, 1,3,3-trimethyl-2-methylene
PubChem CID 8351
IUPAC Name 1,3,3-trimethyl-2-methylideneindole
SMILES CC1(C(=C)N(C2=CC=CC=C21)C)C

Specifications

Melting Point -10°C
Density 0.979
Boiling Point 247°C to 248°C
Flash Point 101°C (213°F)
Refractive Index 1.578
Quantity 50 g
Beilstein 131162
Formula Weight 173.26
Percent Purity 95%
Chemical Name or Material 1,3,3-Trimethyl-2-methyleneindoline
Hazard Category H302-H312-H315-H319-H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 204-235-1
RTECSNumber NM1926500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only