missing translation for 'onlineSavingsMsg'
Learn More

1,3,3-Trimethyl-2-methyleneindoline, 95%

CAS: 118-12-7 | C12H15N | 173.259 g/mol

Supplier:  Thermo Scientific Chemicals A1674130

 View more versions of this product

Catalog No. AAA1674130


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,3,3-Trimethyl-2-methyleneindoline
-10°C
247°C to 248°C
C12H15N
MFCD00005813
131162
ZTUKGBOUHWYFGC-UHFFFAOYSA-N
1,3,3-trimethyl-2-methylideneindole
8351
95%
118-12-7
0.979
101°C (213°F)
1.578
250 g
1,3,3-trimethyl-2-methyleneindoline, fischer's base, fischer base, 1h-indole, 2,3-dihydro-1,3,3-trimethyl-2-methylene, fischer's methylene base, 2-methylene-1,3,3-trimethylindoline, fischers base, ccris 6608, indoline, 2-methylene-1,3,3-trimethyl, indoline, 1,3,3-trimethyl-2-methylene
CC1(C(=C)N(C2=CC=CC=C21)C)C
173.259
173.26

Chemical Identifiers

118-12-7
173.259
ZTUKGBOUHWYFGC-UHFFFAOYSA-N
8351
CC1(C(=C)N(C2=CC=CC=C21)C)C
C12H15N
MFCD00005813
1,3,3-trimethyl-2-methyleneindoline, fischer's base, fischer base, 1h-indole, 2,3-dihydro-1,3,3-trimethyl-2-methylene, fischer's methylene base, 2-methylene-1,3,3-trimethylindoline, fischers base, ccris 6608, indoline, 2-methylene-1,3,3-trimethyl, indoline, 1,3,3-trimethyl-2-methylene
1,3,3-trimethyl-2-methylideneindole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H312-H315-H319-H332

EINECSNumber : 204-235-1

RTECSNumber : NM1926500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only