missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[(3,3-Diphenylpropyl)(methyl)amino]-2-methyl-2-propanol 98.0+%, TCI America™
$76.43 - $203.00
Chemical Identifiers
| CAS | 100442-33-9 |
|---|---|
| Molecular Formula | C20H27NO |
| Molecular Weight (g/mol) | 297.442 |
| MDL Number | MFCD11100703 |
| InChI Key | MQWDISMNBYOLAB-UHFFFAOYSA-N |
| PubChem CID | 10851563 |
| IUPAC Name | 1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-ol |
| SMILES | CC(C)(CN(C)CCC(C1=CC=CC=C1)C2=CC=CC=C2)O |
Chemical Identifiers
| 100442-33-9 | |
| 297.442 | |
| MQWDISMNBYOLAB-UHFFFAOYSA-N | |
| 1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-ol |
| C20H27NO | |
| MFCD11100703 | |
| 10851563 | |
| CC(C)(CN(C)CCC(C1=CC=CC=C1)C2=CC=CC=C2)O |
Specifications
| 100442-33-9 | |
| 145°C | |
| MFCD11100703 | |
| MQWDISMNBYOLAB-UHFFFAOYSA-N | |
| 1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-ol | |
| 10851563 | |
| ≥98.0% (GC,T) | |
| 1-[(3,3-Diphenylpropyl)(methyl)amino]-2-methyl-2-propanol |
| Yellow | |
| C20H27NO | |
| 5 g | |
| CC(C)(CN(C)CCC(C1=CC=CC=C1)C2=CC=CC=C2)O | |
| 297.442 | |
| 297.44 | |
| Liquid |
Safety and Handling
TSCA : No