missing translation for 'onlineSavingsMsg'
Learn More

1-[(2S)-3-(Acetylthio)-2-methylpropionyl]-L-proline Hydrate 90.0+%, TCI America™

Catalog No. a15285g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15285G Supplier TCI America Supplier No. A15285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64838-55-7
Molecular Formula C11H19NO5S
Molecular Weight (g/mol) 277.335
MDL Number MFCD01860852
InChI Key WOOVUTUKNLXXPF-JXLXBRSFSA-N
Synonym s-acetylcaptopril, unii-y139m16fxg, s-1-s-3-acetylthio-2-methylpropanoyl pyrrolidine-2-carboxylic acid, s-3-acetylthio-2-methylpropionyl-l-proline, 1-2s-3-acetylsulfanyl-2-methylpropanoyl-l-proline, n-3-acetylthio-2s-methylpropionyl-l-proline, 1-2s-3-acetylthio-2-methyl-1-oxopropyl-l-proline, 2s-1-2s-3-acetylthio-2-methylpropanoyl pyrrolidine-2-carboxylic acid, 2s-1-3-acetylthio-2-methyl-1-oxopropyl-l-proline, 2s-1-2s-3-acetylsulfanyl-2-methylpropanoyl pyrrolidine-2-carboxylic acid
PubChem CID 16213139
IUPAC Name (2S)-1-[(2S)-3-acetylsulfanyl-2-methylpropanoyl]pyrrolidine-2-carboxylic acid;hydrate
SMILES CC(CSC(=O)C)C(=O)N1CCCC1C(=O)O.O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 259.32
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-[(2S)-3-(Acetylthio)-2-methylpropionyl]-L-proline Hydrate