accessibility menu, dialog, popup

UserName

CH3(S)SH3COON(S)OO

CAS RN 64838-55-7

IUPAC Name: (2S)-1-[(2S)-3-(acetylsulfanyl)-2-methylpropanoyl]pyrrolidine-2-carboxylate
Synonyms: (2S)-1-[(2S)-3-(acetylsulfanyl)-2-methylpropanoyl]pyrrolidine-2-carboxylate
Molecular Weight (g/mol): 258.31
Molecular Formula: C11H16NO4S
InChi Key: ZNQRGUYIKSRYCI-APPZFPTMSA-M
SMILES: C[C@H](CSC(C)=O)C(=O)N1CCC[C@H]1C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results