missing translation for 'onlineSavingsMsg'
Learn More

1(2H)-Phthalazinone, 98+%

CAS: 119-39-1 | C8H6N2O | 146.15 g/mol

$57.34 - $57.34

Chemical Identifiers

CAS 119-39-1
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.15
MDL Number MFCD00006892
InChI Key IJAPPYDYQCXOEF-UHFFFAOYSA-N
Synonym 1 2h-phthalazinone, phthalazin-1 2h-one, phthalazone, phthalazinone, phthalazin-1-one, 1-phthalazinol, phthalazin-1-ol, 1-hydroxyphthalazine, 1 2h phthalazinone, 1,2-dihydrophthalazin-1-one
PubChem CID 8394
ChEBI CHEBI:34023
IUPAC Name 2H-phthalazin-1-one
SMILES C1=CC=C2C(=C1)C=NNC2=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC384570050
SDS View Documents
Thermo Scientific Chemicals
384570050
5 g Glass bottle
Each for $57.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

119-39-1
146.15
IJAPPYDYQCXOEF-UHFFFAOYSA-N
8394
2H-phthalazin-1-one
C8H6N2O
MFCD00006892
1 2h-phthalazinone, phthalazin-1 2h-one, phthalazone, phthalazinone, phthalazin-1-one, 1-phthalazinol, phthalazin-1-ol, 1-hydroxyphthalazine, 1 2h phthalazinone, 1,2-dihydrophthalazin-1-one
CHEBI:34023
C1=CC=C2C(=C1)C=NNC2=O

Specifications

119-39-1
100.0
White to Yellow
98% min. (HPLC)
C8H6N2O
5 g
IJAPPYDYQCXOEF-UHFFFAOYSA-N
2H-phthalazin-1-one
8394
146.15
Crystalline Powder
98.0
184.0°C to 187.0°C
Authentic
Glass bottle
MFCD00006892
1 2h-phthalazinone, phthalazin-1 2h-one, phthalazone, phthalazinone, phthalazin-1-one, 1-phthalazinol, phthalazin-1-ol, 1-hydroxyphthalazine, 1 2h phthalazinone, 1,2-dihydrophthalazin-1-one
C1=CC=C2C(=C1)C=NNC2=O
146.15
CHEBI:34023
98+%
1(2H)-Phthalazinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 204-319-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only