missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 1,25 Dihydroxy Vitamin D3 Competitive ELISA Kit

Catalog No. EEL153
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL153 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL153 Supplier Invitrogen™ Supplier No. EEL153

ELISA

The 1,25 Dihydroxyvitamin D3 (DHVD3) ELISA quantitates DHVD3 in serum, plasma and other biological fluids. Principle of the method The Competitive mulispecies ELISA research use only kit is designed to quantitatively measure 1,25 Dihydroxyvitamin D3independent of species . A 1,25 Dihydroxyvitamin D3 standard is provided to generate a standard curve for the assay and all samples are read off a user generated standard curve.Standards or diluted samples are pipetted into a coated microtiter plate and Biotinylated Detection Ab specific to 1,25 Dihydroxyvitamin D3 is added. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) is added to each well and incubated. Then a TMB substrate solution is added to each well. The enzyme substrate reaction is terminated by the addition of stop solution and the color change is measured on a microplate reader. The concentration of 1,25 Dihydroxyvitamin D3 in the samples is then determined by comparing the OD of the samples to the standard curve. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

1,25-dihydroxy vitamin D3, or calcitriol, is the active form of vitamin D3. It functions as a hormone and plays a crucial role in regulating calcium and phosphorus metabolism. It is mainly localized in the intestines, bones, kidneys, and parathyroid glands. Structurally, it is a steroid hormone derived from cholesterol. Its functions include promoting calcium and phosphorus absorption, maintaining bone health, regulating blood calcium levels, and suppressing parathyroid hormone production.

Specifications

Assay Range 7.81-500 pg/mL
Assay Sensitivity 4.69 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Cell Lysates, Plasma, Serum, Tissue Homogenates
Target Species All Species
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias CalcitriolCalcitriol;
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 50 μL, Plasma 50 μL, Serum 50 μL, Tissue Homogenate 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target 1,25 Dihydroxy Vitamin D3
Test Time 1 hr 20 min
Total Assay Time 2 hr 30 min
Show More Show Less