missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Trimethylsilylethoxy)methyl-1H-pyrazole-5-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog No. p-7029567
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2704303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2704303 Supplier Thermo Scientific Chemicals Supplier No. H2704303
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 903550-12-9
Molecular Formula C15H29BN2O3Si
Molecular Weight (g/mol) 324.303
MDL Number MFCD09037502
InChI Key FVCZMGOMGHMAIX-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-2-trimethylsilyl ethoxy methyl-1h-pyrazole, 1-2-trimethylsilylethoxy methylpyrazole-5-boronic acid, pinacol ester, 1-2-trimethylsilanylethoxymethyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilylethoxy methyl-1h-pyrazole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-2-trimethylsilyl ethoxy methyl pyrazole, 1-2-trimethylsilyl ethoxymethyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilylethoxyl methyl-1h-pyrazole-5-boronic acid pinacol ester, 1-2-trimethylsilyl ethoxymethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1h-pyrazole, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1-2-trimethylsilanyl-ethoxymethyl-1h-pyrazole
PubChem CID 44755201
IUPAC Name trimethyl-[2-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]methoxy]ethyl]silane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=NN2COCC[Si](C)(C)C

Specifications

Quantity 1 g
Formula Weight 324.31
Percent Purity 95%
Chemical Name or Material 1-(2-Trimethylsilylethoxy)methyl-1H-pyrazole-5-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only