accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNNOSiH3CH3CCH3

CAS RN 903550-12-9

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-{[2-(trimethylsilyl)ethoxy]methyl}-1H-pyrazole
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-{[2-(trimethylsilyl)ethoxy]methyl}pyrazole
Molecular Weight (g/mol): 324.3
Molecular Formula: C15H29BN2O3Si
InChi Key: FVCZMGOMGHMAIX-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=NN1COCC[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results