missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Thiazolylazo)-2-naphthol, 98%

Catalog No. AAB2401114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2401114 25 g
AAB2401103 1 g
AAB2401106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2401114 Supplier Thermo Scientific Chemicals Supplier No. B2401114

CAS: 1147-56-4 | C13H9N3OS | 255.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1147-56-4
Molecular Formula C13H9N3OS
Molecular Weight (g/mol) 255.30
MDL Number MFCD00021611
InChI Key GCBAKGGJQPMJLL-QINSGFPZSA-N
Synonym 1-2-thiazolylazo-2-naphthol, 1-2-thiazoylazo-2-naphthol, 2-naphthol, 1-2-thiazolylazo, 2-naphthalenol, 1-2-thiazolylazo, 1z-1-1,3-thiazol-2-ylhydrazinylidene naphthalen-2-one, 1-1,3-thiazol-2-yldiazenyl-2-naphthol, 1z-1-2-thiazolylhydrazinylidene-2-naphthalenone, 1-thiazol-2-ylazo-2-naphthol, 2-naphthalenol, 1-2-2-thiazolyl diazenyl, iomxcgdxeudzak-foclmdbbsa-n
PubChem CID 6308684
SMILES O=C1C=CC2=CC=CC=C2\C1=N\NC1=NC=CS1

Specifications

Melting Point 138°C to 140°C
Quantity 25 g
Beilstein 223783
IUPAC Name (1Z)-1-(1,3-thiazol-2-ylhydrazinylidene)naphthalen-2-one
Formula Weight 255.3
Percent Purity 98%
Chemical Name or Material 1-(2-Thiazolylazo)-2-naphthol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-555-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only