missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Thiazolyl)piperazine 98.0+%, TCI America™

Catalog No. T3273200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3273200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3273200MG Supplier TCI America Supplier No. T3273200MG
Only null left

Chemical Identifiers

CAS 42270-37-1
Molecular Formula C7H11N3S
Molecular Weight (g/mol) 169.25
MDL Number MFCD00778419
InChI Key WQFWIVTXNKRNJZ-UHFFFAOYSA-N
Synonym 2-(1-Piperazinyl)thiazole
PubChem CID 911806
IUPAC Name 1-(1,3-thiazol-2-yl)piperazine
SMILES C1CN(CCN1)C1=NC=CS1

Specifications

Color Yellow
Boiling Point 125°C
Quantity 200 mg
Formula Weight 169.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(2-Thiazolyl)piperazine