missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Pyridin-3-ylphenyl)methanamine dihydrochloride, ≥97%, Thermo Scientific™

Chemical Identifiers

CAS 859833-18-4
Molecular Formula C12H14Cl2N2
Molecular Weight (g/mol) 257.158
MDL Number MFCD08060476
InChI Key SSFBEGXFELCUJT-UHFFFAOYSA-N
Synonym 1-2-pyridin-3-ylphenyl methanamine dihydrochloride, 1-2-pyridin-3-ylphenyl methylamine dihydrochloride, 2-pyridin-3-yl phenyl methanamine dihydrochloride, 1-2-pyridin-3-ylphenyl methanaminedihydrochloride, 1-2-pyridin-3-yl phenyl methanamine dihydrochloride, 2-pyridin-3-yl-benzylamine dihydrochloride, 2-3-pyridinyl benzenemethanamine dihydrochloride, 2-3-pyridyl phenyl methylamine, chloride, chloride
PubChem CID 18525754
IUPAC Name (2-pyridin-3-ylphenyl)methanamine;dihydrochloride
SMILES C1=CC=C(C(=C1)CN)C2=CN=CC=C2.Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC41863CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC41863DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

859833-18-4
257.158
SSFBEGXFELCUJT-UHFFFAOYSA-N
18525754
C1=CC=C(C(=C1)CN)C2=CN=CC=C2.Cl.Cl
C12H14Cl2N2
MFCD08060476
1-2-pyridin-3-ylphenyl methanamine dihydrochloride, 1-2-pyridin-3-ylphenyl methylamine dihydrochloride, 2-pyridin-3-yl phenyl methanamine dihydrochloride, 1-2-pyridin-3-ylphenyl methanaminedihydrochloride, 1-2-pyridin-3-yl phenyl methanamine dihydrochloride, 2-pyridin-3-yl-benzylamine dihydrochloride, 2-3-pyridinyl benzenemethanamine dihydrochloride, 2-3-pyridyl phenyl methylamine, chloride, chloride
(2-pyridin-3-ylphenyl)methanamine;dihydrochloride

Specifications

859833-18-4
100.0
C12H14Cl2N2
1-2-pyridin-3-ylphenyl methanamine dihydrochloride, 1-2-pyridin-3-ylphenyl methylamine dihydrochloride, 2-pyridin-3-yl phenyl methanamine dihydrochloride, 1-2-pyridin-3-ylphenyl methanaminedihydrochloride, 1-2-pyridin-3-yl phenyl methanamine dihydrochloride, 2-pyridin-3-yl-benzylamine dihydrochloride, 2-3-pyridinyl benzenemethanamine dihydrochloride, 2-3-pyridyl phenyl methylamine, chloride, chloride
C1=CC=C(C(=C1)CN)C2=CN=CC=C2.Cl.Cl
257.158
257.16
1-(2-Pyridin-3-ylphenyl)methanamine dihydrochloride
96.5
97%
MFCD08060476
SSFBEGXFELCUJT-UHFFFAOYSA-N
(2-pyridin-3-ylphenyl)methanamine;dihydrochloride
18525754
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents