Learn More
1-(2-Pyridin-3-ylphenyl)methanamine dihydrochloride, ≥97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC41863DA
| Quantity | 1g |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 859833-18-4 | |
| 257.158 | |
| SSFBEGXFELCUJT-UHFFFAOYSA-N | |
| 18525754 | |
| C1=CC=C(C(=C1)CN)C2=CN=CC=C2.Cl.Cl |
| C12H14Cl2N2 | |
| MFCD08060476 | |
| 1-2-pyridin-3-ylphenyl methanamine dihydrochloride, 1-2-pyridin-3-ylphenyl methylamine dihydrochloride, 2-pyridin-3-yl phenyl methanamine dihydrochloride, 1-2-pyridin-3-ylphenyl methanaminedihydrochloride, 1-2-pyridin-3-yl phenyl methanamine dihydrochloride, 2-pyridin-3-yl-benzylamine dihydrochloride, 2-3-pyridinyl benzenemethanamine dihydrochloride, 2-3-pyridyl phenyl methylamine, chloride, chloride | |
| (2-pyridin-3-ylphenyl)methanamine;dihydrochloride |
Specifications
| 1-(2-Pyridin-3-ylphenyl)methanamine dihydrochloride | |
| 96.5 | |
| 97% | |
| C12H14Cl2N2 | |
| MFCD08060476 | |
| SSFBEGXFELCUJT-UHFFFAOYSA-N | |
| (2-pyridin-3-ylphenyl)methanamine;dihydrochloride | |
| 18525754 | |
| 97% |
| 859833-18-4 | |
| 100.0 | |
| Amber Glass Bottle | |
| 1g | |
| 1-2-pyridin-3-ylphenyl methanamine dihydrochloride, 1-2-pyridin-3-ylphenyl methylamine dihydrochloride, 2-pyridin-3-yl phenyl methanamine dihydrochloride, 1-2-pyridin-3-ylphenyl methanaminedihydrochloride, 1-2-pyridin-3-yl phenyl methanamine dihydrochloride, 2-pyridin-3-yl-benzylamine dihydrochloride, 2-3-pyridinyl benzenemethanamine dihydrochloride, 2-3-pyridyl phenyl methylamine, chloride, chloride | |
| C1=CC=C(C(=C1)CN)C2=CN=CC=C2.Cl.Cl | |
| 257.158 | |
| 257.16 |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.