missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Propynyloxy)naphthalene 95.0+%, TCI America™

Catalog No. P22275G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22275G 5 g
P22271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22275G Supplier TCI America Supplier No. P22275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18542-45-5
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.222
MDL Number MFCD03787730
InChI Key SQUJYDFTMPTTLT-UHFFFAOYSA-N
Synonym 1-(Propargyloxy)naphthalene
PubChem CID 29132
IUPAC Name 1-prop-2-ynoxynaphthalene
SMILES C#CCOC1=CC=CC2=CC=CC=C21

Specifications

Color Yellow
Boiling Point 75°C
Quantity 5 g
Formula Weight 182.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Propynyloxy)naphthalene
RTECSNumber QK0377500
TSCA No
Recommended Storage Refrigerator