accessibility menu, dialog, popup

UserName

CAS RN 18542-45-5

CHO

CAS RN 18542-45-5

IUPAC Name: 1-(prop-2-yn-1-yloxy)naphthalene
Synonyms: 1-(prop-2-yn-1-yloxy)naphthalene
Molecular Weight (g/mol): 182.22
Molecular Formula: C13H10O
InChi Key: SQUJYDFTMPTTLT-UHFFFAOYSA-N
SMILES: C#CCOC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results