missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[(2-Propynyloxy)methyl]pyrene 98.0+%, TCI America™
Supplier: TCI America P22261G
Specifications
| 1-[(2-Propynyloxy)methyl]pyrene | |
| 85°C | |
| C20H14O | |
| 1-[(Propargyloxy)methyl]pyrene | |
| C#CCOCC1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1 | |
| 270.331 | |
| 270.33 | |
| Crystalline Powder |
| 1115084-83-7 | |
| Yellow | |
| 1 g | |
| PHLBHJYVJNAPAY-UHFFFAOYSA-N | |
| 1-(prop-2-ynoxymethyl)pyrene | |
| 91972178 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1115084-83-7 | |
| 270.331 | |
| 1-[(Propargyloxy)methyl]pyrene | |
| 1-(prop-2-ynoxymethyl)pyrene |
| C20H14O | |
| PHLBHJYVJNAPAY-UHFFFAOYSA-N | |
| 91972178 | |
| C#CCOCC1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1 |
Safety and Handling
TSCA : No