missing translation for 'onlineSavingsMsg'
Learn More

1-[(2-Propynyloxy)methyl]pyrene 98.0+%, TCI America™

Catalog No. P2226200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2226200MG 200 mg
P22261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2226200MG Supplier TCI America Supplier No. P2226200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1115084-83-7
Molecular Formula C20H14O
Molecular Weight (g/mol) 270.331
InChI Key PHLBHJYVJNAPAY-UHFFFAOYSA-N
Synonym 1-[(Propargyloxy)methyl]pyrene
PubChem CID 91972178
IUPAC Name 1-(prop-2-ynoxymethyl)pyrene
SMILES C#CCOCC1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1

Specifications

Melting Point 85°C
Color Yellow
Quantity 200 mg
Formula Weight 270.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-[(2-Propynyloxy)methyl]pyrene