missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Propynyl)piperazine 97.0+%, TCI America™
Supplier: TCI America P2228200MG
Specifications
| 1-(2-Propynyl)piperazine | |
| 60°C | |
| 90°C | |
| MFCD08445213 | |
| 1-Propargylpiperazine | |
| C#CCN1CCNCC1 | |
| 124.187 | |
| 124.19 | |
| Crystalline Powder |
| 52070-67-4 | |
| Yellow | |
| C7H12N2 | |
| 200 mg | |
| GWCSATTUAOHJDK-UHFFFAOYSA-N | |
| 1-prop-2-ynylpiperazine | |
| 14627369 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 52070-67-4 | |
| 124.187 | |
| GWCSATTUAOHJDK-UHFFFAOYSA-N | |
| 14627369 | |
| C#CCN1CCNCC1 |
| C7H12N2 | |
| MFCD08445213 | |
| 1-Propargylpiperazine | |
| 1-prop-2-ynylpiperazine |
Safety and Handling
TSCA : No