missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Propynyl)piperazine 97.0+%, TCI America™
$229.29 - $682.24
Chemical Identifiers
| CAS | 52070-67-4 |
|---|---|
| Molecular Formula | C7H12N2 |
| Molecular Weight (g/mol) | 124.187 |
| MDL Number | MFCD08445213 |
| InChI Key | GWCSATTUAOHJDK-UHFFFAOYSA-N |
| Synonym | 1-Propargylpiperazine |
| PubChem CID | 14627369 |
| IUPAC Name | 1-prop-2-ynylpiperazine |
| SMILES | C#CCN1CCNCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P2228200MG
|
TCI America
P2228200MG |
200 mg |
Each for $229.29
|
|
|||||
|
P22281G
|
TCI America
P22281G |
1 g |
Each for $682.24
|
|
|||||
Chemical Identifiers
| 52070-67-4 | |
| 124.187 | |
| GWCSATTUAOHJDK-UHFFFAOYSA-N | |
| 14627369 | |
| C#CCN1CCNCC1 |
| C7H12N2 | |
| MFCD08445213 | |
| 1-Propargylpiperazine | |
| 1-prop-2-ynylpiperazine |
Specifications
| 52070-67-4 | |
| Yellow | |
| C7H12N2 | |
| 200 mg | |
| GWCSATTUAOHJDK-UHFFFAOYSA-N | |
| 1-prop-2-ynylpiperazine | |
| 14627369 | |
| ≥97.0% (GC,T) | |
| 1-(2-Propynyl)piperazine |
| 60°C | |
| 90°C | |
| MFCD08445213 | |
| 1-Propargylpiperazine | |
| C#CCN1CCNCC1 | |
| 124.187 | |
| 124.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No