missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Propanediol, 99%, extra pure
CAS: 57-55-6 | C3H8O2 | 76.09 g/mol
Supplier: Thermo Scientific Chemicals 158720025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1, 2-Propanediol | |
| 98.5 | |
| -60.0°C | |
| 187.0°C | |
| Authentic | |
| Glass Bottle | |
| 1.4310 to 1.4330 | |
| MFCD00064272 | |
| 01, 472 | |
| 15, 7968 | |
| Solubility in water: completely soluble. Other solubilities: soluble in ether, miscible with acetone and chloroform | |
| CC(CO)O | |
| 76.09 | |
| 45 mPa.s (20°C) | |
| 76.09 | |
| Extra Pure |
| 57-55-6 | |
| 100.0 | |
| 1.0300g/mL | |
| 99°C | |
| 98.5% min. (GC) | |
| C3H8O2 | |
| CH3CH(OH)CH2OH | |
| 2.5 L | |
| 1.03 | |
| 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
| DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
| propane-1,2-diol | |
| 1030 | |
| CHEBI:16997 | |
| 99% | |
| Viscous Liquid |
Chemical Identifiers
| 57-55-6 | |
| 76.09 | |
| DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
| 1030 | |
| propane-1,2-diol |
| C3H8O2 | |
| MFCD00064272 | |
| 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
| CHEBI:16997 | |
| CC(CO)O |
Safety and Handling
EINECSNumber : 200-338-
RUO – Research Use Only