missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanediol, 99%, extra pure

Catalog No. p-3598236
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158720010 1 L Glass Bottle
AC158720025 2.5 L Glass Bottle
AC158720250 25 L Metal drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158720010 Supplier Thermo Scientific Chemicals Supplier No. 158720010
Only null left

CAS: 57-55-6 | C3H8O2 | 76.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.09
MDL Number MFCD00064272
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
PubChem CID 1030
ChEBI CHEBI:16997
IUPAC Name propane-1,2-diol
SMILES CC(CO)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -60.0°C
Density 1.0300g/mL
Boiling Point 187.0°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4310 to 1.4330
Linear Formula CH3CH(OH)CH2OH
Quantity 1 L
Beilstein 01, 472
Specific Gravity 1.03
Merck Index 15, 7968
Solubility Information Solubility in water: completely soluble. Other solubilities: soluble in ether, miscible with acetone and chloroform
Viscosity 45 mPa.s (20°C)
Formula Weight 76.09
Percent Purity 99%
Grade Extra Pure
Physical Form Viscous Liquid
Chemical Name or Material 1, 2-Propanediol
Show More Show Less
EINECSNumber 200-338-

RUO – Research Use Only