missing translation for 'onlineSavingsMsg'
Learn More

1,2-Propanediol, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187373
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014715 1 mL
111014716 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014715 Supplier Merck Emd Millipore Supplier No. 122791MLF
Only null left
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 57-55-6
Ignition Point 779°F
Vapor Pressure 0.08 mmHg (20°C)
Vapor Density 2.62 (vs. air)
Density 1.036 g/mL (at 25°C)
Flash Point 103°C
Physical Form Neat
Linear Formula CH3 CH(OH)CH2 OH
MDL Number MFCD00064272
Melting Point -60°C
Boiling Point 187°C
Synonym 1,2-PDO; Propylene glycol
Type Analytical Standard
Quantity 1 mL
Formula Weight 76.09
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less