missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Phenylethyl)-4-(phenylethynyl)benzene, 97%

Catalog No. AAH3005803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3005803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3005803 Supplier Thermo Scientific Chemicals Supplier No. H3005803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 906650-60-0 | C22H18 | 282.386 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 906650-60-0
Molecular Formula C22H18
Molecular Weight (g/mol) 282.386
MDL Number MFCD12407075
InChI Key GXNVEXGZHSEHNG-UHFFFAOYSA-N
Synonym 1-2-phenylethyl-4-phenylethynyl benzene, 1-2-phenylethyl-4-2-phenylethynyl benzene, 4-phenylethynylbibenzyl, 4-phenethyldiphenylacetylene, 4-4-phenethylphenyl ethynyl benzene
PubChem CID 20724073
IUPAC Name 1-(2-phenylethyl)-4-(2-phenylethynyl)benzene
SMILES C1=CC=C(C=C1)CCC2=CC=C(C=C2)C#CC3=CC=CC=C3

Specifications

Melting Point 104°C to 106°C
Quantity 1 g
Formula Weight 282.39
Percent Purity 97%
Chemical Name or Material 1-(2-Phenylethyl)-4-(phenylethynyl)benzene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only