CAS RN 906650-60-0
IUPAC Name:
1-(2-phenylethyl)-4-(2-phenylethynyl)benzene
Synonyms:
1-(2-phenylethyl)-4-(2-phenylethynyl)benzene
Molecular Weight (g/mol):
282.39
Molecular Formula:
C22H18
InChi Key:
GXNVEXGZHSEHNG-UHFFFAOYSA-N
SMILES:
C(CC1=CC=C(C=C1)C#CC1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
1-(2-Phenylethyl)-4-(phenylethynyl)benzene, 97%
1-(2-Phenylethyl)-4-(phenylethynyl)benzene, 97%
CAS: 906650-60-0 Molecular Formula: C22H18 Molecular Weight (g/mol): 282.386 MDL Number: MFCD12407075 InChI Key: GXNVEXGZHSEHNG-UHFFFAOYSA-N Synonym: 1-2-phenylethyl-4-phenylethynyl benzene,1-2-phenylethyl-4-2-phenylethynyl benzene,4-phenylethynylbibenzyl,4-phenethyldiphenylacetylene,4-4-phenethylphenyl ethynyl benzene PubChem CID: 20724073 IUPAC Name: 1-(2-phenylethyl)-4-(2-phenylethynyl)benzene SMILES: C1=CC=C(C=C1)CCC2=CC=C(C=C2)C#CC3=CC=CC=C3