missing translation for 'onlineSavingsMsg'
Learn More

1,2-Phenylenediamine 98.0+%, TCI America™

Catalog No. p-7134465
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P016825G 25 g
P0168100G 100 g
P0168500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P016825G Supplier TCI America Supplier No. P016825G

Chemical Identifiers

CAS 95-54-5
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00007721
InChI Key GEYOCULIXLDCMW-UHFFFAOYSA-N
Synonym o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine
PubChem CID 7243
ChEBI CHEBI:34043
IUPAC Name benzene-1,2-diamine
SMILES C1=CC=C(C(=C1)N)N

Specifications

Melting Point 103°C
Boiling Point 265°C
Quantity 25 g
UN Number 1673
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Phenylenediamine
EINECSNumber (3)-0185
RTECSNumber SS7875000
TSCA Yes