missing translation for 'onlineSavingsMsg'
Learn More

1,2-Phenylenediamine 98.0+%, TCI America™

Catalog No. p096825g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
P096825G 25 g
1 options

Catalog No. P096825G

Supplier: TCI America P096825G

Only null left

Chemical Identifiers

CAS 95-54-5
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00007721
InChI Key GEYOCULIXLDCMW-UHFFFAOYSA-N
Synonym o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine
PubChem CID 7243
ChEBI CHEBI:34043
IUPAC Name benzene-1,2-diamine
SMILES C1=CC=C(C(=C1)N)N

Specifications

Melting Point 103°C
Color White
Boiling Point 265°C
Quantity 25 g
UN Number 1673
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Phenylenediamine
EINECSNumber (3)-0185
RTECSNumber SS7875000
TSCA Yes
Recommended Storage Refrigerator