missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Phenylenediamine 98.0+%, TCI America™
$86.64 - $246.43
Chemical Identifiers
| CAS | 95-54-5 |
|---|---|
| Molecular Formula | C6H8N2 |
| Molecular Weight (g/mol) | 108.144 |
| MDL Number | MFCD00007721 |
| InChI Key | GEYOCULIXLDCMW-UHFFFAOYSA-N |
| Synonym | o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine |
| PubChem CID | 7243 |
| ChEBI | CHEBI:34043 |
| IUPAC Name | benzene-1,2-diamine |
| SMILES | C1=CC=C(C(=C1)N)N |
Chemical Identifiers
| 95-54-5 | |
| 108.144 | |
| GEYOCULIXLDCMW-UHFFFAOYSA-N | |
| 7243 | |
| benzene-1,2-diamine |
| C6H8N2 | |
| MFCD00007721 | |
| o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine | |
| CHEBI:34043 | |
| C1=CC=C(C(=C1)N)N |
Specifications
| 95-54-5 | |
| White | |
| C6H8N2 | |
| 1 g | |
| o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine | |
| C1=CC=C(C(=C1)N)N | |
| 108.144 | |
| CHEBI:34043 | |
| ≥98.0% (GC,T) | |
| 1,2-Phenylenediamine [for Biochemical Research] |
| 103°C | |
| 265°C | |
| MFCD00007721 | |
| 1673 | |
| GEYOCULIXLDCMW-UHFFFAOYSA-N | |
| benzene-1,2-diamine | |
| 7243 | |
| 108.14 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0185
RTECSNumber : SS7875000
TSCA : Yes