missing translation for 'onlineSavingsMsg'
Learn More

1,2-Phenylenediamine Dihydrochloride 98.0+%, TCI America™

Catalog No. p017125g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
P017125G 25 g
P0171500G 500 g
2 options

Catalog No. P017125G

Supplier: TCI America P017125G

Only null left

Chemical Identifiers

CAS 615-28-1
Molecular Formula C6H10Cl2N2
Molecular Weight (g/mol) 181.06
MDL Number MFCD00012966
InChI Key RIIWUGSYXOBDMC-UHFFFAOYSA-N
Synonym 1,2-phenylenediamine dihydrochloride, o-phenylenediamine dihydrochloride, 1,2-benzenediamine, dihydrochloride, opd easy-tablets, opd tablets, benzene-1,2-diamine dihydrochloride, usaf ek-678, o-phenylenediamine.2hcl, 1,2-benzenediamine dihydrochloride, 1,2-diaminobenzene dihydrochloride
PubChem CID 11990
IUPAC Name benzene-1,2-diamine;dihydrochloride
SMILES C1=CC=C(C(=C1)N)N.Cl.Cl

Specifications

Melting Point 205°C
Color Red-White
Quantity 25 g
UN Number 2811
Formula Weight 181.06
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Phenylenediamine Dihydrochloride
RTECSNumber ST0175000
TSCA Yes