missing translation for 'onlineSavingsMsg'
Learn More

1-[2-Oxo-2-(2-pyridyl)ethyl]pyridinium Iodide 98.0+%, TCI America™

Catalog No. O04381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04381G 1 g
O04385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04381G Supplier TCI America Supplier No. O04381G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26482-00-8
Molecular Formula C12H11IN2O
Molecular Weight (g/mol) 326.137
MDL Number MFCD01690884
InChI Key BWAWQRQYQUSQEV-UHFFFAOYSA-M
PubChem CID 213499
IUPAC Name 2-pyridin-1-ium-1-yl-1-pyridin-2-ylethanone;iodide
SMILES C1=CC=[N+](C=C1)CC(=O)C2=CC=CC=N2.[I-]

Specifications

Melting Point 190°C
Quantity 1 g
Formula Weight 326.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-[2-Oxo-2-(2-pyridyl)ethyl]pyridinium Iodide
RTECSNumber UU6897000
TSCA No
Recommended Storage Refrigerator