accessibility menu, dialog, popup

UserName

CAS RN 26482-00-8

IUPAC Name: 1-[2-oxo-2-(pyridin-2-yl)ethyl]pyridin-1-ium iodide
Synonyms: 1-[2-oxo-2-(pyridin-2-yl)ethyl]pyridin-1-ium iodide
Molecular Weight (g/mol): 326.14
Molecular Formula: C12H11IN2O
InChi Key: BWAWQRQYQUSQEV-UHFFFAOYSA-M
SMILES: [I-].O=C(C[N+]1=CC=CC=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results