missing translation for 'onlineSavingsMsg'
Learn More

1,2-Naphthalic Anhydride 98.0+%, TCI America™

Catalog No. N05835G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05835G 5 g
N058325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05835G Supplier TCI America Supplier No. N05835G

Chemical Identifiers

CAS 5343-99-7
Molecular Formula C12H6O3
Molecular Weight (g/mol) 198.177
MDL Number MFCD00191561
InChI Key IDVDAZFXGGNIDQ-UHFFFAOYSA-N
Synonym 1,2-Naphthalenedicarboxylic Anhydride
PubChem CID 21437
IUPAC Name benzo[e][2]benzofuran-1,3-dione
SMILES C1=CC=C2C(=C1)C=CC3=C2C(=O)OC3=O

Specifications

Melting Point 170°C
Quantity 5 g
Formula Weight 198.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Naphthalic Anhydride