accessibility menu, dialog, popup

UserName

OOO

CAS RN 5343-99-7

IUPAC Name: 1H,3H-naphtho[1,2-c]furan-1,3-dione
Synonyms: naphtho[1,2-c]furan-1,3-dione
Molecular Weight (g/mol): 198.18
Molecular Formula: C12H6O3
InChi Key: IDVDAZFXGGNIDQ-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=C3C=CC=CC3=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results