missing translation for 'onlineSavingsMsg'
Learn More

1,2-Naphthalenedicarbonitrile 98.0+%, TCI America™

Catalog No. N05785G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05785G 5 g
N05781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05785G Supplier TCI America Supplier No. N05785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19291-76-0
Molecular Formula C12H6N2
Molecular Weight (g/mol) 178.19
MDL Number MFCD00191673
InChI Key ZNPWYAMBOPRTHW-UHFFFAOYSA-N
Synonym 1,2-Dicyanonaphthalene
PubChem CID 350412
IUPAC Name naphthalene-1,2-dicarbonitrile
SMILES N#CC1=CC=C2C=CC=CC2=C1C#N

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 178.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Naphthalenedicarbonitrile