accessibility menu, dialog, popup

UserName

NN

CAS RN 19291-76-0

IUPAC Name: naphthalene-1,2-dicarbonitrile
Synonyms: naphthalene-1,2-dicarbonitrile
Molecular Weight (g/mol): 178.19
Molecular Formula: C12H6N2
InChi Key: ZNPWYAMBOPRTHW-UHFFFAOYSA-N
SMILES: N#CC1=CC=C2C=CC=CC2=C1C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results