missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(morpholin-4-ylmethyl)phenyl]methanamine, 97%, Thermo Scientific™

Catalog No. p-4510702
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC48613DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. CC48613DA missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' CC48613DA

Chemical Identifiers

CAS 91271-82-8
Molecular Formula C12H18N2O
Molecular Weight (g/mol) 206.289
InChI Key NMFAEZHWSZZJOA-UHFFFAOYSA-N
Synonym 2-morpholin-4-ylmethylbenzylamine, 1-2-morpholin-4-ylmethyl phenyl methanamine, 2-morpholin-4-ylmethyl-benzylamine, 1-2-morpholin-4-ylmethyl phenyl methylamine, 2-morpholinomethyl phenyl methanamine, 2-morpholin-4-ylmethyl phenyl methanamine, 2-morpholin-4-ylmethyl phenyl methylamine, 2-morpholin-4-ylmethylphenylamine, 2-morpholin-4-ylmethyl benzyl amine
PubChem CID 4962390
IUPAC Name [2-(morpholin-4-ylmethyl)phenyl]methanamine
SMILES C1COCCN1CC2=CC=CC=C2CN

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 206.29
Percent Purity 95%
Chemical Name or Material 1-[2-(Morpholin-4-ylmethyl)phenyl]methanamine