missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Methylphenyl)piperazine, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 48.0°C to 52.0°C |
| Color | White to Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Quantity | 1 g |
| Formula Weight | 176.26 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Harmful if swallowed. Harmful in contact with skin. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water |
| EINECSNumber | 254-481-9 |
RUO – Research Use Only