missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methyl-5-nitrophenyl)guanidine Nitrate 97.0+%, TCI America™

Catalog No. p-7133323
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26131G 1 g
M26135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26131G Supplier TCI America Supplier No. M26131G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 152460-08-7
Molecular Formula C8H11N5O5
Molecular Weight (g/mol) 257.206
MDL Number MFCD09842424
InChI Key IINMQQJNRFDBMV-UHFFFAOYSA-N
PubChem CID 11528991
IUPAC Name 2-(2-methyl-5-nitrophenyl)guanidine;nitric acid
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])N=C(N)N.[N+](=O)(O)[O-]

Specifications

Melting Point 218°C
Color Yellow
Quantity 1 g
Formula Weight 257.21
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Methyl-5-nitrophenyl)guanidine Nitrate