missing translation for 'onlineSavingsMsg'
Learn More

1,2-Indanedione, 95%

Catalog No. AC460920010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460920010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC460920010 Supplier Thermo Scientific Chemicals Supplier No. 460920010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16214-27-0 | C9H6O2 | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16214-27-0
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.15
InChI Key WFFZGYRTVIPBFN-UHFFFAOYSA-N
Synonym 1h-indene-1,2 3h-dione, 1,2-indanedione, indan-1,2-dione, 1,2-indandione, indandione, 2,3-dihydro-1h-indene-1,2-dione, indane dione, acmc-20a7vb, d01xgp
PubChem CID 123358
IUPAC Name 3H-indene-1,2-dione
SMILES C1C2=CC=CC=C2C(=O)C1=O

Specifications

Melting Point 117.0°C to 124.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 146.15
Percent Purity 95%
Chemical Name or Material 1,2-Indanedione
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. If eye irritation persists: Get medical advice/attention. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only