accessibility menu, dialog, popup

UserName

OO

CAS RN 16214-27-0

IUPAC Name: 2,3-dihydro-1H-indene-1,2-dione
Synonyms: 3H-indene-1,2-dione
Molecular Weight (g/mol): 146.15
Molecular Formula: C9H6O2
InChi Key: WFFZGYRTVIPBFN-UHFFFAOYSA-N
SMILES: O=C1CC2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)