missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Hydroxyethyl)-3-methylimidazolium Chloride 98.0+%, TCI America™
$115.57 - $346.71
Chemical Identifiers
| CAS | 61755-34-8 |
|---|---|
| Molecular Formula | C6H11ClN2O |
| MDL Number | MFCD12761437 |
Chemical Identifiers
| 61755-34-8 | |
| C6H11ClN2O | |
| MFCD12761437 |
Specifications
| 61755-34-8 | |
| White-Yellow | |
| MFCD12761437 | |
| 162.62 | |
| Crystalline Powder |
| 83°C | |
| C6H11ClN2O | |
| 5 g | |
| ≥98.0% (HPLC) | |
| 1-(2-Hydroxyethyl)-3-methylimidazolium Chloride |
Safety and Handling
TSCA : No