missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Hydroxyethyl)-3-methylimidazolium Chloride 98.0+%, TCI America™
Supplier: TCI America H125425G
Specifications
| 1-(2-Hydroxyethyl)-3-methylimidazolium Chloride | |
| 83°C | |
| C6H11ClN2O | |
| 25 g | |
| ≥98.0% (HPLC) |
| 61755-34-8 | |
| White-Yellow | |
| MFCD12761437 | |
| 162.62 | |
| Crystalline Powder |
Chemical Identifiers
| 61755-34-8 | |
| C6H11ClN2O | |
| MFCD12761437 |
Safety and Handling
TSCA : No