missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Hexanediol 96.0+%, TCI America™
$70.25 - $510.72
Chemical Identifiers
| CAS | 6920-22-5 |
|---|---|
| Molecular Formula | C6H14O2 |
| Molecular Weight (g/mol) | 118.18 |
| MDL Number | MFCD00010737 |
| InChI Key | FHKSXSQHXQEMOK-LURJTMIESA-N |
| Synonym | 1,2-hexanediol, 1,2-dihydroxyhexane, dl-1,2-hexanediol, 1,2-hexane diol, dl-hexane-1,2-diol, 5,6-dihydroxyhexane, 1,2-hexylene glycol, acmc-209o6j, 2,2,6,6-tetrakis hydroxymet cyclohexanol |
| PubChem CID | 94335 |
| IUPAC Name | (2S)-hexane-1,2-diol |
| SMILES | CCCC[C@H](O)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
H068825ML
|
N/A | 25 mL |
Each for $70.25
|
|
|||||
|
H0688100ML
|
N/A | 100 mL |
Each for $186.07
|
|
|||||
|
H0688500ML
|
N/A | 500 mL |
Each for $510.72
|
|
|||||
Chemical Identifiers
| C6H14O2 | |
| MFCD00010737 | |
| 1,2-hexanediol, 1,2-dihydroxyhexane, dl-1,2-hexanediol, 1,2-hexane diol, dl-hexane-1,2-diol, 5,6-dihydroxyhexane, 1,2-hexylene glycol, acmc-209o6j, 2,2,6,6-tetrakis hydroxymet cyclohexanol | |
| (2S)-hexane-1,2-diol |
Specifications
| 6920-22-5 | |
| 118°C | |
| MFCD00010737 | |
| FHKSXSQHXQEMOK-LURJTMIESA-N | |
| (2S)-hexane-1,2-diol | |
| 94335 | |
| ≥96.0% (GC) | |
| 1,2-Hexanediol |
| Colorless | |
| C6H14O2 | |
| 1,2-hexanediol, 1,2-dihydroxyhexane, dl-1,2-hexanediol, 1,2-hexane diol, dl-hexane-1,2-diol, 5,6-dihydroxyhexane, 1,2-hexylene glycol, acmc-209o6j, 2,2,6,6-tetrakis hydroxymet cyclohexanol | |
| CCCC[C@H](O)CO | |
| 118.18 | |
| 118.18 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0240
TSCA : Yes