missing translation for 'onlineSavingsMsg'
Learn More

1,2-Hexanediol 96.0+%, TCI America™

Catalog No. H068825ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H068825ML 25 mL
H0688100ML 100 mL
H0688500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H068825ML Supplier TCI America Supplier No. H068825ML

Chemical Identifiers

CAS 6920-22-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00010737
InChI Key FHKSXSQHXQEMOK-LURJTMIESA-N
Synonym 1,2-hexanediol, 1,2-dihydroxyhexane, dl-1,2-hexanediol, 1,2-hexane diol, dl-hexane-1,2-diol, 5,6-dihydroxyhexane, 1,2-hexylene glycol, acmc-209o6j, 2,2,6,6-tetrakis hydroxymet cyclohexanol
PubChem CID 94335
IUPAC Name (2S)-hexane-1,2-diol
SMILES CCCC[C@H](O)CO

Specifications

Color Colorless
Boiling Point 118°C
Quantity 25 mL
Formula Weight 118.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Hexanediol
EINECSNumber (2)-0240
TSCA Yes