missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluorophenyl)piperazine 98.0+%, TCI America™

Catalog No. F072825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F072825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F072825G Supplier TCI America Supplier No. F072825G
Only null left

Chemical Identifiers

CAS 1011-15-0
Molecular Formula C10H13FN2
Molecular Weight (g/mol) 180.226
MDL Number MFCD00040726
InChI Key IVTZRJKKXSKXKO-UHFFFAOYSA-N
Synonym 1-2-fluorophenyl piperazine, 1-2-fluorophenyl-piperazine, 1-2-fluoro-phenyl-piperazine, piperazine, 1-2-fluorophenyl, chembl9429, 2-fluorophenyl piperazine, ofpp, pubchem8573, 1-2-fluorophenyl piperazinehydrochloride, 2-fluorophenylpiperazine
PubChem CID 70529
IUPAC Name 1-(2-fluorophenyl)piperazine
SMILES C1CN(CCN1)C2=CC=CC=C2F

Specifications

Melting Point 49°C
Color White
Boiling Point 93°C
Quantity 25 g
Formula Weight 180.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystals
Chemical Name or Material 1-(2-Fluorophenyl)piperazine