CAS RN 1011-15-0
IUPAC Name:
1-(2-fluorophenyl)piperazine
Synonyms:
1-(2-fluorophenyl)piperazine
Molecular Weight (g/mol):
180.23
Molecular Formula:
C10H13FN2
InChi Key:
IVTZRJKKXSKXKO-UHFFFAOYSA-N
SMILES:
FC1=CC=CC=C1N1CCNCC1
1
–
1
of
1
results
1
1-(2-Fluorophenyl)piperazine 98.0+%, TCI America™
CAS: 1011-15-0 Molecular Formula: C10H13FN2 Molecular Weight (g/mol): 180.226 MDL Number: MFCD00040726 InChI Key: IVTZRJKKXSKXKO-UHFFFAOYSA-N Synonym: 1-2-fluorophenyl piperazine,1-2-fluorophenyl-piperazine,1-2-fluoro-phenyl-piperazine,piperazine, 1-2-fluorophenyl,chembl9429,2-fluorophenyl piperazine,ofpp,pubchem8573,1-2-fluorophenyl piperazinehydrochloride,2-fluorophenylpiperazine PubChem CID: 70529 IUPAC Name: 1-(2-fluorophenyl)piperazine SMILES: C1CN(CCN1)C2=CC=CC=C2F