missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluorobenzoyl)pyrrolidine, 97%, Thermo Scientific™

Catalog No. AAH56339MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56339MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH56339MD Supplier Thermo Scientific Chemicals Supplier No. H56339MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2‘-fluoro-6-pyrrol-2-phenyl-4-quinolone, a derivative of 1-(2-Fluorobenzoyl)pyrrolidine exhibited the most potent cytotoxic activities against renal and melanoma tumor cell lines.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 347909-09-5
Molecular Formula C11H12FNO
Molecular Weight (g/mol) 193.221
MDL Number MFCD00460952
InChI Key AZBXHPRYPHBYCV-UHFFFAOYSA-N
Synonym 1-2-fluorobenzoyl pyrrolidine, 2-fluorophenyl 1-pyrrolidinyl methanone, 2-fluorophenyl-pyrrolidin-1-ylmethanone
PubChem CID 3497148
IUPAC Name (2-fluorophenyl)-pyrrolidin-1-ylmethanone
SMILES C1CCN(C1)C(=O)C2=CC=CC=C2F

Specifications

Quantity 250 mg
Formula Weight 193.22
Percent Purity 97%
Chemical Name or Material 1-(2-Fluorobenzoyl)pyrrolidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures