missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxydecane 97.0+%, TCI America™

Catalog No. p-7128930
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
E031525ML 25 mL
E0315500ML 500 mL
2 options

Catalog No. E031525ML

Supplier: TCI America E031525ML

Only null left

Chemical Identifiers

CAS 2404-44-6
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00005158
InChI Key AAMHBRRZYSORSH-UHFFFAOYNA-N
Synonym 1,2-epoxydecane, oxirane, octyl, octyloxirane, oxirane, 2-octyl, 1,2-decylene oxide, ccris 2615, epoxides, c>8-alkyl, decene oxide, epoxy decane, oxirane,octyl
PubChem CID 16993
IUPAC Name 2-octyloxirane
SMILES CCCCCCCCC1CO1

Specifications

Color Yellow
Boiling Point 208°C
Quantity 25 mL
Formula Weight 156.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxydecane
EINECSNumber (2)-0220
TSCA Yes