missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxydecane, 97%

CAS: 2404-44-6 | C10H20O | 156.27 g/mol

Supplier:  Thermo Scientific Chemicals L0728906

 View more versions of this product

Catalog No. AAL0728906


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,2-Epoxydecane
0.84
78°C (172°F)
1.429
5 g
1,2-epoxydecane, oxirane, octyl, octyloxirane, oxirane, 2-octyl, 1,2-decylene oxide, ccris 2615, epoxides, c>8-alkyl, decene oxide, epoxy decane, oxirane,octyl
CCCCCCCCC1CO1
156.27
156.27
2404-44-6
92°C to 93°C (14 mmHg)
C10H20O
MFCD00005158
104132
AAMHBRRZYSORSH-UHFFFAOYNA-N
2-octyloxirane
16993
97%

Chemical Identifiers

2404-44-6
156.27
AAMHBRRZYSORSH-UHFFFAOYNA-N
16993
CCCCCCCCC1CO1
C10H20O
MFCD00005158
1,2-epoxydecane, oxirane, octyl, octyloxirane, oxirane, 2-octyl, 1,2-decylene oxide, ccris 2615, epoxides, c>8-alkyl, decene oxide, epoxy decane, oxirane,octyl
2-octyloxirane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 219-295-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only