missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxydecane, 97%

Catalog No. AAL0728914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0728914 25 g
AAL0728906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0728914 Supplier Thermo Scientific Chemicals Supplier No. L0728914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2404-44-6 | C10H20O | 156.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2404-44-6
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00005158
InChI Key AAMHBRRZYSORSH-UHFFFAOYNA-N
Synonym 1,2-epoxydecane, oxirane, octyl, octyloxirane, oxirane, 2-octyl, 1,2-decylene oxide, ccris 2615, epoxides, c>8-alkyl, decene oxide, epoxy decane, oxirane,octyl
PubChem CID 16993
IUPAC Name 2-octyloxirane
SMILES CCCCCCCCC1CO1

Specifications

Density 0.84
Boiling Point 92°C to 93°C (14 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.429
Quantity 25 g
Beilstein 104132
Formula Weight 156.27
Percent Purity 97%
Chemical Name or Material 1,2-Epoxydecane
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 219-295-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only